Dipropyl phthalate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
B24056.09
B24056.18
B24056.30
B24056.09EA
1690
CZK
B24056.09
B24056.18
B24056.30
Dipropyl phthalate ≥98%
Dipropyl phthalate
Purity: 98%
CAS No.: 131-16-8
Molecular Formula: C14H18O4
Formula Weight: 250.30
Flash Point:
Vzorec:
C₁₄H₁₈O₄ MW: 250,29 g/mol Bod varu: 168…170 °C (3 mmHg) Hustota: 1,082 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009371 CAS číslo: 131-16-8 EINECS: 205-015-8 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...