Tisk…

Saccharin ≥98%

Dodavatel: Thermo Fisher Scientific

B23938.0E B23938.22 B23938.36
B23938.0EEA 12740 CZK
B23938.0E B23938.22 B23938.36
Saccharin ≥98%
Saccharin

Purity: 98+% CAS No.: 81-07-2 Molecular Formula: C7H5NO3S Formula Weight: 183.18 Melting Point: 227-230° Beilstein Reference: 6888 Merck Reference: 14,8311 MDL No.: MFCD00005866

Vzorec: C₇H₅NO₃S
Bod tání: 227…230 °C
Hustota: 0,828
Storage Temperature: Ambient
MDL Number: MFCD00005866
CAS číslo: 81-07-2
EINECS: 201-321-0
Merck Index: 14,8311

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR