Tisk…

Diethyl terephthalate 95%

Dodavatel: Thermo Fisher Scientific

Synonyms: Diethyl benzene-1,4-dicarboxylate

B21401.09 B21401.18
B21401.09EA 1600 CZK
B21401.09 B21401.18
Diethyl terephthalate 95%
Diethyl terephthalate

Purity: 95% CAS No.: 636-09-9 Molecular Formula: C12H14O4 Formula Weight: 222.24 Melting Point: 44-46° MDL No.: MFCD00039891

Vzorec: C₁₂H₁₄O₄
Bod tání: 44…46 °C
Hustota: 1,100
Storage Temperature: Ambient
MDL Number: MFCD00039891
CAS číslo: 636-09-9
EINECS: 211-249-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR