2-[2-(Hexyloxy)ethoxy]ethan-1-ol ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B20774.0D
B20774.0F
B20774.AP
B20774.0DEA
24510
CZK
B20774.0D
B20774.0F
B20774.AP
2-[2-(Hexyloxy)ethoxy]ethan-1-ol ≥97%
2-[2-(Hexyloxy)ethoxy]ethan-1-ol
Purity: 97%
CAS No.: 112-59-4
Molecular Formula: C10H22O3
Formula Weight: 190.28
Flash Point: 123°(253°F)
Refractive Index: 1.4370
Melting Point: -40°
Boiling Point: 259-261°
Beilstein Reference: 1743959
MDL No.: MFCD00010703
Vzorec:
C₁₀H₂₂O₃ Bod varu: 259…261 °C Bod tání: –40 °C Hustota: 0,932 Bod vzplanutí: 123 °C (253 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010703 CAS číslo: 112-59-4 EINECS: 203-988-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...