2,4,6-Triphenylpyrilium tetrafluoroborate ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2,4,6-Triphenylpyrylium tetrafluoroborate
A18217.06
A18217.14
A18217.22
A18217.06EA
2040
CZK
A18217.06
A18217.14
A18217.22
2,4,6-Triphenylpyrilium tetrafluoroborate ≥97%
2,4,6-Triphenylpyrilium tetrafluoroborate
Purity: 97%
CAS No.: 448-61-3
Molecular Formula: C23H17BF4O
Formula Weight: 396.20
Melting Point: 248-254°
Beilstein Reference: 3586533
Vzorec:
C₂₃H₁₇BF₄O MW: 396,19 g/mol Bod tání: 248…254 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012001 CAS číslo: 448-61-3 EINECS: 207-186-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...