Tisk…

Menthone (mixture of isomers) ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Menthone, 2-Isopropyl-5-Methylcyclohexanone

A17665.0B A17665.18 A17665.30
A17665.0BEA 12530 CZK
A17665.0B A17665.18 A17665.30
Menthone (mixture of isomers) ≥98%
Menthone (mixture of isomers)

Purity: 98% CAS No.: 10458-14-7 Molecular Formula: C10H18O Formula Weight: 154.25 Flash Point: 72°(162°F) Boiling Point: ca 207° MDL No.: MFCD00062998

Vzorec: C₁₀H₁₈O
MW: 154,25 g/mol
Bod varu: ∼207 °C
Hustota: 0,894
Bod vzplanutí: 72 °C (162 °F)
Storage Temperature: Ambient
MDL Number: MFCD00062998
CAS číslo: 10458-14-7
EINECS: 233-944-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR