Tisk…

Pyridinium tribromide tech. 90%, technická kvalita

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A15684.14 A15684.22 A15684.36
A15684.14EA 1500 CZK
A15684.14 A15684.22 A15684.36
Pyridinium tribromide tech. 90%, technická kvalita
Pyridinium tribromide

Purity: 90% CAS No.: 39416-48-3 Molecular Formula: C5H5N.HBr3 Formula Weight: 319.82 Melting Point: 128-136° Beilstein Reference: 3690144 Merck Reference: 14,7973 UN No.: 3261 MDL No.: MFCD00013223

Vzorec: C₅H₆Br₃N
Bod tání: 128…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013223
CAS číslo: 39416-48-3
EINECS: 254-446-8
UN: 3261
ADR: 8,III
Merck Index: 14,7973

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR