Chloro(dimethyl)(phenyl)silane 97+%, rods
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Chlorodimethylphenylsilane
A15638.06
A15638.14
A15638.22
A15638.06EA
1700
CZK
A15638.06
A15638.14
A15638.22
Chloro(dimethyl)(phenyl)silane 97+%, rods
Chloro(dimethyl)(phenyl)silane
Purity: 97+%
CAS No.: 768-33-2
Molecular Formula: C8H11ClSi
Formula Weight: 170.72
Flash Point: 72°(161°F)
Refractive Index: 1.5090
Boiling Point: 192-193°
Beilstein Reference: 606292
UN No.: 2987
MDL No.: MFCD00000499
Vzorec:
C₈H₁₁ClSi MW: 170.71 Bod varu: 192…193 °C Hustota: 1.032 Bod vzplanutí: 72 °C (161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000499 CAS číslo: 768-33-2 EINECS: 212-193-0 UN: 2987 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...