Tisk…

Diheptyl phthalate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A15466.09 A15466.18
A15466.09EA 2120 CZK
A15466.09 A15466.18
Diheptyl phthalate ≥98%
Diheptyl phthalate

Purity: 98+% CAS No.: 3648-21-3 Molecular Formula: C22H34O4 Formula Weight: 362.51 Flash Point: 213°(415°F) Refractive Index: 1.4860 Boiling Point: 360° Beilstein Reference: 2336623 MDL No.: MFCD00043680

Vzorec: C₂₂H₃₄O₄
Bod varu: 360 °C
Hustota: 0,992
Bod vzplanutí: 213 °C (415 °F)
Storage Temperature: Ambient
MDL Number: MFCD00043680
CAS číslo: 3648-21-3
EINECS: 222-885-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR