Tisk…

2-Bromoterephthalic acid ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A14393.06 A14393.14 A14393.22
A14393.06EA 2010 CZK
A14393.06 A14393.14 A14393.22
2-Bromoterephthalic acid ≥97%
2-Bromoterephthalic acid

Purity: 97% CAS No.: 586-35-6 Molecular Formula: C8H5BrO4 Formula Weight: 245.03 Melting Point: 297-300° Beilstein Reference: 2616163 UN No.: 2811 MDL No.: MFCD00002403

Vzorec: C₈H₅BrO₄
MW: 245.03
Bod tání: 297…300 °C
Storage Temperature: Ambient
MDL Number: MFCD00002403
CAS číslo: 586-35-6
EINECS: 209-572-8
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR