Tisk…

Homophthalic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A14297.18 A14297.30
A14297.18EA 1740 CZK
A14297.18 A14297.30
Homophthalic acid ≥99%
Homophthalic acid

Purity: 99% CAS No.: 89-51-0 Molecular Formula: C9H8O4 Formula Weight: 180.16 Melting Point: ca 180° dec. Beilstein Reference: 1872069

Vzorec: C₉H₈O₄
Storage Temperature: Ambient
MDL Number: MFCD00004326
CAS číslo: 89-51-0
EINECS: 201-913-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR