Methyl-2-furoate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 98+%
CAS No.: 611-13-2
Molecular Formula: C6H6O3
Formula Weight: 126.11
Flash Point: 73°(163°F)
Refractive Index: 1.4870
Boiling Point: 180-181°
Beilstein Reference: 111110
Merck Reference: 14,4307
UN No.: 2810
MDL No.: MFCD00003236
Vzorec:
C₆H₆O₃ Bod varu: 180…181 °C Hustota: 1,179 Bod vzplanutí: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003236 CAS číslo: 611-13-2 EINECS: 210-254-6 UN: 2810 ADR: 6.1,III Merck Index: 14,4307 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...