Tisk…

2-Furohydrazide ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-Furoic acid hydrazide, 2-Furoylhydrazine, 2-Furancarboxylic acid Hydrazide, 2-Furoic hydrazide, 2-Furoic acid Hydrazide, 2-Furancarboxylic acid hydrazide, 2-Furoic Hydrazide

A13630.09 A13630.18
A13630.09EA 1580 CZK
A13630.09 A13630.18
2-Furohydrazide ≥98%
2-Furohydrazide

Purity: 98% CAS No.: 3326-71-4 Molecular Formula: C5H6N2O2 Formula Weight: 126.12 Melting Point: 77-81° Beilstein Reference: 114435

Vzorec: C₅H₆N₂O₂
MW: 126,11 g/mol
Bod tání: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00003235
CAS číslo: 3326-71-4
EINECS: 222-046-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR