Tisk…

Ethyl-3-bromopropionate ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Ethyl 3-bromopropanoate

A12402.14 A12402.22 A12402.36 AB148814.0025
A12402.14EA 1580 CZK
A12402.14 A12402.22 A12402.36 ABCRAB148814.0025
Ethyl-3-bromopropionate ≥99%
Ethyl-3-bromopropionate

Purity: 99% CAS No.: 539-74-2 Molecular Formula: BrCH2CH2CO2CH2CH3 Formula Weight: 181.03 Flash Point: 79°(174°F) Refractive Index: 1.4520 Boiling Point: 67-71°/10mm Beilstein Reference: 1700933 MDL No.: MFCD00000251

Vzorec: C₅H₉BrO₂
Bod varu: 67…71 °C (10 mmHg)
Hustota: 1,416
Bod vzplanutí: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000251
CAS číslo: 539-74-2
EINECS: 208-724-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR