2-Chloroethyl toluene-4-sulphonate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Chloroethyl p-toluenesulphonate
A11863.14
A11863.22
A11863.36
A11863.14EA
1880
CZK
A11863.14
A11863.22
A11863.36
2-Chloroethyl toluene-4-sulphonate ≥97%
2-Chloroethyl toluene-4-sulphonate
Purity: 97%
CAS No.: 80-41-1
Molecular Formula: C9H11ClO3S
Formula Weight: 234.70
Storage Temperature: 2°C to 8°C
Flash Point:
Vzorec:
C₉H₁₁ClO₃S Bod varu: 158…159 °C (0,5 mmHg) Hustota: 1,294 Bod vzplanutí: >110 °C (230 °F) |
MDL Number:
MFCD00000970 CAS číslo: 80-41-1 EINECS: 201-277-2 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...