Pyridine-2-carbaldehyde ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Pyridinecarboxaldehyde, 2-Pyridinecarbaldehyde, Pyridine-2-carboxaldehyde
A11391.14
A11391.22
A11391.36
A11391.14EA
1020
CZK
A11391.14
A11391.22
A11391.36
Pyridine-2-carbaldehyde ≥99%
Pyridine-2-carbaldehyde
Purity: 99%
CAS No.: 1121-60-4
Molecular Formula: C6H5NO
Formula Weight: 107.11
Storage Temperature: 2°C to 8°C
Flash Point: 68°(154°F)
Refractive Index: 1.5360
Odour: Characteristic
pH: 6.5
Melting Point: -21°
Boiling Point: 180-181°
Beilstein Reference: 105341
UN No.: 2810
MDL No.: MFCD00006290
Vzorec:
C₆H₅NO MW: 107,11 g/mol Bod varu: 180…181 °C Bod tání: –21 °C Hustota: 1,124 Bod vzplanutí: 68 °C (154 °F) |
MDL Number:
MFCD00006290 CAS číslo: 1121-60-4 EINECS: 214-333-6 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...