Tisk…

Methyl-3,5-dibromo-4-hydroxybenzoate ≥98%

Dodavatel: Thermo Scientific
Varování

A10799.09 A10799.18 A10799.30
A10799.09EA 841 CZK
A10799.09 A10799.18 A10799.30
Methyl-3,5-dibromo-4-hydroxybenzoate ≥98%
Methyl-3,5-dibromo-4-hydroxybenzoate

Purity: 98% CAS No.: 41727-47-3 Molecular Formula: C8H6Br2O3 Formula Weight: 309.94 Melting Point: 123-125° Beilstein Reference: 2373854

Vzorec: C₈H₆Br₂O₃
MW: 309,94 g/mol
Bod tání: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00016416
CAS číslo: 41727-47-3
EINECS: 255-521-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR