Dimethyl-2-aminoterephthalate 99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate
A10567.0B
A10567.18
A10567.30
A10567.0BEA
21070
CZK
A10567.0B
A10567.18
A10567.30
Dimethyl-2-aminoterephthalate 99%
Dimethyl-2-aminoterephthalate
Purity: 99%
CAS No.: 5372-81-6
Molecular Formula: C10H11NO4
Formula Weight: 209.20
Melting Point: 130-134°
Beilstein Reference: 2808505
Vzorec:
C₁₀H₁₁NO₄ MW: 209,2 g/mol Bod tání: 130…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008427 CAS číslo: 5372-81-6 EINECS: 226-364-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...