Tisk…

Gallium(III) ethanolate

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Gallium(III) ethoxide

44859.03 44859.06
44859.03EA 2480 CZK
44859.03 44859.06
Gallium(III) ethanolate
Gallium(III) ethanolate

CAS No.: 2572-25-0 Molecular Formula: C6H15GaO3.xHOCH2CH3 Formula Weight: 204.90 Flash Point: 68°(154°F) Physical Form: Liquid Density: 1.25 Appearance: Yellow MDL No.: MFCD01866730

Vzorec: C₆H₁₅GaO₃
Hustota: 1,25
Bod vzplanutí: 68 °C (154 °F)
Storage Temperature: Ambient
MDL Number: MFCD01866730
CAS číslo: 2572-25-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR