Tisk…

Iron(III) ethoxide ≥99.6% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování


Packaged under argon.

41863.02 41863.04 41863.09 41863.18
41863.02EA 1350 CZK
41863.02 41863.04 41863.09 41863.18
Iron(III) ethoxide ≥99.6% (metals basis)
Iron(III) ethoxide

Purity: 99.6% CAS No.: 5058-42-4 Molecular Formula: C6H15FeO3 Formula Weight: 191.03 Physical Form: Powder/Chunk, Packaged under argon Sensitivity: Moisture Sensitive Appearance: Brown to dark grey Odour: Odourless Melting Point: 120° Boiling Point: 155°/0.1mm Solubility: Soluble in anhydrous hexane, alcohols UN No.: 3181 MDL No.: MFCD00078028


Precursor for Sol-Gel processes

Vzorec: C₆H₁₅FeO₃
Bod varu: 155 °C (0,1 mmHg)
Bod tání: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00078028
CAS číslo: 5058-42-4
EINECS: 225-760-2
UN: 3181
ADR: 4.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR