Tisk…
Yttrium(III) trifluoromethanesulphonate hydrate
Dodavatel: Thermo Fisher Scientific
Varování
40315.04 40315.09
40315.04EA 1230 CZK
40315.04 40315.09
Yttrium(III) trifluoromethanesulphonate hydrate
Yttrium(III) trifluoromethanesulphonate hydrate
CAS No.: 34629-25-9 Molecular Formula: Y(OSO2CF3)3.xH2O Formula Weight: 536.10(anhy) Physical Form: Powder MDL No.: MFCD00209623
Vzorec: C₃F₉O₉S₃Y·H₂O
MW: 554.13
Storage Temperature: Ambient
MDL Number: MFCD00209623
CAS číslo: 34629-25-9