Tisk…
Terbium(III) trifluoromethanesulphonate ≥98%
Dodavatel: Thermo Fisher Scientific
Varování
40312.04 40312.09
40312.04EA 3030 CZK
40312.04 40312.09
Terbium(III) trifluoromethanesulphonate ≥98%
Terbium(III) trifluoromethanesulphonate
Purity: 98% CAS No.: 148980-31-8 Molecular Formula: Tb(OSO2CF3)3 Formula Weight: 606.12 Physical Form: Powder
Vzorec: C₃F₉O₉S₃Tb
MW: 606.14
Storage Temperature: Ambient
MDL Number: MFCD00192461
CAS číslo: 148980-31-8