Iron(II) methoxide ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Packaged under argon.
22552.04
22552.09
22552.04EA
3880
CZK
22552.04
22552.09
Iron(II) methoxide ≥98%
Iron(II) methoxide
Purity: 98%
CAS No.: 7245-21-8
Molecular Formula: C2H6FeO2
Formula Weight: 117.92
Physical Form: Powder, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Green
Odour: Alcohol-like
UN No.: 3181
MDL No.: MFCD00061474
Vzorec:
C₂H₆FeO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00061474 CAS číslo: 7245-21-8 UN: 3181 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...