Tantalum(V) methanolate
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Tantalum(V) methoxide
18839.06
18839.14
18839.06EA
5980
CZK
18839.06
18839.14
Tantalum(V) methanolate
Tantalum(V) methanolate
CAS No.: 865-35-0
Molecular Formula: C5H15O5Ta
Formula Weight: 336.12
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Melting Point: 50°
Boiling Point: 189°/10mm
UN No.: 3181
MDL No.: MFCD00058817
Vzorec:
C₅H₁₅O₅Ta MW: 336.12 Bod varu: 189 °C (10 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00058817 CAS číslo: 865-35-0 UN: 3181 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...