Tisk…

(±)-α-Terpineol ≥96%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: α-Terpineol, Terpineol, 2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol

16285.22 16285.36 16285.14
16285.22EA 1900 CZK
16285.22 16285.36 16285.14
(±)-α-Terpineol ≥96%
(±)-α-Terpineol

Purity: 96% CAS No.: 98-55-5 Molecular Formula: C10H18O Formula Weight: 154.24 Flash Point: 90°(194°F) Physical Form: Liquid Density: 0.930 Refractive Index: 1.4830 Appearance: Colourless to pale yellow Melting Point: 31-32° Boiling Point: 210-219° Merck Reference: 14,9171 MDL No.: MFCD00001557


A naturally-occuring monoterpene alcohol

Vzorec: C₁₀H₁₈O
Bod varu: 210…219 °C
Bod tání: 31…32 °C
Hustota: 0,930
Bod vzplanutí: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001557
CAS číslo: 98-55-5
EINECS: 202-680-6
Merck Index: 14,9171

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR