Tantalum(V) ethanolate ≥99.999% (metals basis) (max. 100 ppm Nb)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Tantalum(V) ethoxide
14643.03
14643.09
14643.18
14643.03EA
2180
CZK
14643.03
14643.09
14643.18
Tantalum(V) ethanolate ≥99.999% (metals basis) (max. 100 ppm Nb)
Tantalum(V) ethanolate
Purity: 99.999%
CAS No.: 6074-84-6
Molecular Formula: C10H25O5Ta
Formula Weight: 406.25
Flash Point: 30°(87°F)
Physical Form: Liquid
Density: 1.566
Refractive Index: 1.4880
Sensitivity: Air and Moisture Sensitive
Melting Point: 18-19°
Boiling Point: 145°/0.1mm
Solubility: Decomposes in water. Soluble in organic solvents
Beilstein Reference: 3678999
UN No.: 2924
MDL No.: MFCD00049785
Vzorec:
C₁₀H₂₅O₅Ta Bod varu: 145 °C (0,1 mmHg) Bod tání: 18…19 °C Hustota: 1,566 Bod vzplanutí: 30 °C (87 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00049785 CAS číslo: 6074-84-6 EINECS: 228-010-2 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...