Tisk…

Silver perchlorate monohydrate 99,9 Metals basis

Dodavatel: Thermo Fisher Scientific
Nebezpečí

11624.04 11624.09 11624.18
11624.04EA 1080 CZK
11624.04 11624.09 11624.18
Silver perchlorate monohydrate 99,9 Metals basis
Silver perchlorate monohydrate

Purity: 99.9% CAS No.: 14242-05-8 Molecular Formula: AgClO4.H2O Formula Weight: 225.34 (207.32anhy) Physical Form: Crystalline Density: 2.800 Appearance: White Odour: Odourless Melting Point: 485° dec. Merck Reference: 14,8523 UN No.: 1481 MDL No.: MFCD00149128

Vzorec: AgClO₄·1H₂O
MW: 225.33
Bod tání: 485 °C (rozkl.)
Hustota: 2.800
Storage Temperature: Ambient
MDL Number: MFCD00149128
CAS číslo: 14242-05-8
EINECS: 232-035-4
UN: 1481
ADR: 5.1 II
Merck Index: 14,8523

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR