Aluminium trifluoromethanesulphonate 99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Aluminum trifluoromethanesulphonate, Aluminum triflate, Aluminum tris(trifluoromethanesulphonate)
383260100
383260500
ACRO383260100EA
3980
CZK
ACRO383260100
ACRO383260500
Aluminium trifluoromethanesulphonate 99%
Aluminium trifluoromethanesulphonate
Vzorec:
C₃AlF₉O₉S₃ MW: 474.19 Bod tání: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143596 CAS číslo: 74974-61-1 UN: 3261 ADR: 8 III |
Výsledky specifikačního testu
Appearance | White Powder |
Infrared spectrum | Conforms |
Titration Complexometric | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...