Diethylaluminium chloride 0.9 M in toluene, AcroSeal®
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
DEAC, Diethylaluminum chloride
381151000
381158000
381151000.EA
2330
CZK
381151000.
381158000.
Diethylaluminium chloride 0.9 M in toluene, AcroSeal®
Diethylaluminium chloride
Vzorec:
C₄H₁₀AlCl MW: 120.56 Bod varu: 208 Bod tání: -50 Hustota: 0.961 Bod vzplanutí: 4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000459 CAS číslo: 96-10-6 EINECS: 202-477-2 UN: 3399 ADR: 4.3 I |
Výsledky specifikačního testu
Appearance | Clear colorless Liquid |
Strength | 11.5 to 12.5 % (on Al basis) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...