Tisk…

Chloro(dimethyl)(phenyl)silane 95%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Chlorodimethylphenylsilane

151070250 151070050 151071000
ACRO151070250EA 3470 CZK
ACRO151070250 ACRO151070050 ACRO151071000
Chloro(dimethyl)(phenyl)silane 95%
Chloro(dimethyl)(phenyl)silane
Vzorec: C₈H₁₁ClSi
MW: 170,71 g/mol
Bod varu: 80…84 °C (16 mmHg)
Hustota: 1,032 g/cm³
Bod vzplanutí: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00000499
CAS číslo: 768-33-2
EINECS: 212-193-0
UN: 2987
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5060 to 1.5100 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR