Tisk…

Pyridine-2-carbaldehyde 99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-Pyridinecarboxaldehyde, 2-Pyridinecarbaldehyde, Pyridine-2-carboxaldehyde

131820250 131821000 131825000
ACRO131820250EA 1080 CZK
ACRO131820250 ACRO131821000 ACRO131825000
Pyridine-2-carbaldehyde 99%
Pyridine-2-carbaldehyde
Vzorec: C₆H₅NO
MW: 107,11 g/mol
Bod varu: 180…182 °C (761 mmHg)
Hustota: 1,121 g/cm³
Bod vzplanutí: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006290
CAS číslo: 1121-60-4
EINECS: 214-333-6
UN: 2810
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance Clear yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5343 to 1.5373 (20°C, 589 nm)
GC ≥99.5 %
UV at 220 nm A: ≤0.7
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Carbonyl compounds ≤0.03 % (as C2H5CHO)
Isopropyl alcohol ≤0.05 %
Titrable acid 0.0004 meq/g
E/E ≤0.01 %
Methanol ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR