Tisk…

Cyclopentylamine 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Aminocyclopentane

L01966.14 L01966.22
L01966.14EA 1090 CZK
L01966.14 L01966.22
Cyclopentylamine 99%
Cyclopentylamine

Purity: 99% CAS No.: 1003-03-8 Molecular Formula: C5H11N Formula Weight: 85.15 Flash Point: 12°(53°F) Refractive Index: 1.4505 Odour: Amine-like pH: 12.5 Melting Point: -85° Boiling Point: 106-108° Beilstein Reference: 635706 UN No.: 3286 MDL No.: MFCD00001380

Vzorec: C₅H₁₁N
Bod varu: 106…108 °C
Bod tání: –85 °C
Hustota: 0,862
Bod vzplanutí: 12 °C (53 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001380
CAS číslo: 1003-03-8
EINECS: 213-697-3
UN: 3286
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR