Tisk…

Cyclobutylamine 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Cyclobutylamine, Cyclobutanamine, Aminocyclobutane

A13423.03 A13423.06 A13423.14
A13423.03EA 1930 CZK
A13423.03 A13423.06 A13423.14
Cyclobutylamine 98%
Cyclobutylamine

Purity: 98% CAS No.: 2516-34-9 Molecular Formula: C4H9N Formula Weight: 71.12 Flash Point: -4°(24°F) Refractive Index: 1.4376 Boiling Point: 82-83° Beilstein Reference: 2069297 UN No.: 2733 MDL No.: MFCD00001328

Vzorec: C₄H₉N
MW: 71,12 g/mol
Bod varu: 82…83 °C
Hustota: 0,836
Bod vzplanutí: –4 °C (24 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001328
CAS číslo: 2516-34-9
EINECS: 219-736-0
UN: 2733
ADR: 3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR