Cyclobutylamine 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Cyclobutylamine, Cyclobutanamine, Aminocyclobutane
209110010
209110050
ACRO209110010EA
2030
CZK
ACRO209110010
ACRO209110050
Cyclobutylamine 98%
Cyclobutylamine
Vzorec:
C₄H₉N MW: 71,12 g/mol Bod varu: 81,4 °C (1013 hPa) Hustota: 0,83 g/cm³ Bod vzplanutí: –4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001328 CAS číslo: 2516-34-9 EINECS: 219-736-0 UN: 2733 ADR: 3,II |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4360 to 1.4400 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...