Tisk…

1-Aminocyclohexanecarboxylic acid 98%

Dodavatel: Thermo Scientific

Synonyms: 1-Amino-1-cyclohexanecarboxylic acid, 1-Aminocyclohexane-1-carboxylic acid

A11853.06 A11853.14
A11853.06EA 1190 CZK
A11853.06 A11853.14
1-Aminocyclohexanecarboxylic acid 98%
1-Aminocyclohexanecarboxylic acid

Purity: 98% CAS No.: 2756-85-6 Molecular Formula: C7H13NO2 Formula Weight: 143.19 Melting Point: 300° Beilstein Reference: 2355692

Vzorec: C₇H₁₃NO₂
MW: 143,19 g/mol
Bod varu: 272 °C (1013 hPa)
Bod tání: >300 °C
Hustota: 1,229 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001487
CAS číslo: 2756-85-6
EINECS: 220-411-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR