Cyclobutanecarboxylic acid ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A14480.06
A14480.14
A14480.22
A14480.06EA
1010
CZK
A14480.06
A14480.14
A14480.22
Cyclobutanecarboxylic acid ≥98%
Cyclobutanecarboxylic acid
Purity: 98+%
CAS No.: 3721-95-7
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 83°(181°F)
Refractive Index: 1.4433
Odour: Stench
Melting Point: -20°
Boiling Point: 194-195°
Beilstein Reference: 1816777
UN No.: 3265
MDL No.: MFCD00001323
Vzorec:
C₅H₈O₂ MW: 100,12 g/mol Bod varu: 194…195 °C Bod tání: –20 °C Hustota: 1,055 Bod vzplanutí: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001323 CAS číslo: 3721-95-7 EINECS: 223-072-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...