1,4-Cyclohexanedione ≥98%
Dodavatel: Thermo Scientific
A11100.14
A11100.22
A11100.36
A11100.14EA
2150
CZK
A11100.14
A11100.22
A11100.36
1,4-Cyclohexanedione ≥98%
1,4-Cyclohexanedione
Purity: 98%
CAS No.: 637-88-7
Molecular Formula: C6H8O2
Formula Weight: 112.13
Storage Temperature: 2°C to 8°C
Flash Point: 132°(269°F)
Melting Point: 77-78°
Boiling Point: 132°/20mm
Beilstein Reference: 774152
MDL No.: MFCD00001606
Vzorec:
C₆H₈O₂ MW: 112,13 g/mol Bod varu: 132 °C (20 mmHg) Bod tání: 77…78 °C Hustota: 1,28 g/cm³ (20 °C) Bod vzplanutí: 132 °C (269 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001606 CAS číslo: 637-88-7 EINECS: 211-306-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...