Tisk…

1,3-Cyclopentanedione ≥98%

Dodavatel: Thermo Scientific

Synonyms: Cyclopentane-1,3-dione

L10336.03 L10336.06
L10336.03EA 1730 CZK
L10336.03 L10336.06
1,3-Cyclopentanedione ≥98%
1,3-Cyclopentanedione

Purity: 98% CAS No.: 3859-41-4 Molecular Formula: C5H6O2 Formula Weight: 98.10 Melting Point: 151-153° Beilstein Reference: 1362728

Vzorec: C₅H₆O₂
MW: 98,1 g/mol
Bod tání: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00001405
CAS číslo: 3859-41-4
EINECS: 223-372-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR