Cyclooctanone ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B20911.14
B20911.22
B20911.36
B20911.14EA
1100
CZK
B20911.14
B20911.22
B20911.36
Cyclooctanone ≥98%
Cyclooctanone
Purity: 98%
CAS No.: 502-49-8
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 72°(161°F)
Odour: Pleasant
Boiling Point: 195-197°
Beilstein Reference: 1280738
UN No.: 1759
MDL No.: MFCD00001754
Vzorec:
C₈H₁₄O Bod varu: 195…197 °C Bod tání: 37…44 °C Hustota: 0,960 Bod vzplanutí: 72 °C (161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001754 CAS číslo: 502-49-8 EINECS: 207-940-2 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...