Tisk…

Bromocyclopentane ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Cyclopentyl bromide

A15455.22 A15455.36
A15455.22EA 1200 CZK
A15455.22 A15455.36
Bromocyclopentane ≥98%
Bromocyclopentane

Purity: 98+% CAS No.: 137-43-9 Molecular Formula: C5H9Br Formula Weight: 149.04 Flash Point: 42°(107°F) Refractive Index: 1.4881 Odour: Odourless Boiling Point: 137-139° Beilstein Reference: 1209256 UN No.: 1993 MDL No.: MFCD00001359

Vzorec: C₅H₉Br
Bod varu: 137…139 °C
Hustota: 1,390
Bod vzplanutí: 42 °C (107 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001359
CAS číslo: 137-43-9
EINECS: 205-294-6
UN: 1993
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR