Bromocycloheptane ≥97%
Dodavatel: Thermo Scientific
Varování
|
Synonyms:
Cycloheptyl bromide
B23110.06
B23110.14
B23110.06EA
927
CZK
B23110.06
B23110.14
Bromocycloheptane ≥97%
Bromocycloheptane
Purity: 97%
CAS No.: 2404-35-5
Molecular Formula: C7H13Br
Formula Weight: 177.09
Flash Point: 68°(154°F)
Refractive Index: 1.5052
Boiling Point: 71-72°/10mm
Beilstein Reference: 1098389
MDL No.: MFCD00004149
Vzorec:
C₇H₁₃Br MW: 177,08 g/mol Bod varu: 71…72 °C (10 mmHg) Hustota: 1,300 Bod vzplanutí: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004149 CAS číslo: 2404-35-5 EINECS: 219-294-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...