Bromocycloheptane 97%
Dodavatel: Thermo Scientific
Synonyms:
Cycloheptyl bromide
111080250
111081000
ACRO111080250EA
3040
CZK
ACRO111080250
ACRO111081000
Bromocycloheptane 97%
Bromocycloheptane
Vzorec:
C₇H₁₃Br MW: 177,08 g/mol Bod varu: 71…72 °C (10 mmHg) Hustota: 1,28 g/cm³ Bod vzplanutí: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004149 CAS číslo: 2404-35-5 EINECS: 219-294-9 |
Výsledky specifikačního testu
Appearance | Clear light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5035 to 1.5055 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...