2-Methyl-8-quinolinol ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
8-Hydroxy-2-methylquinoline, 8-Hydroxyquinaldine, 2-Methyloxine
L06989.22
L06989.36
L06989.22EA
1580
CZK
L06989.22
L06989.36
2-Methyl-8-quinolinol ≥98%
2-Methyl-8-quinolinol
Purity: 98%
CAS No.: 826-81-3
Molecular Formula: C10H9NO
Formula Weight: 159.19
Melting Point: 70-74°
Boiling Point: 267°
UN No.: 3077
MDL No.: MFCD00006765
Vzorec:
C₁₀H₉NO MW: 159,19 g/mol Bod varu: 267 °C Bod tání: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006765 CAS číslo: 826-81-3 EINECS: 212-562-6 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...