4-Quinolinol 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Quinolin-4-ol, 4-Hydroxyquinoline
121890100
121890500
ACRO121890100EA
2810
CZK
ACRO121890100
ACRO121890500
4-Quinolinol 98%
4-Quinolinol
Vzorec:
C₉H₇NO MW: 145,16 g/mol Bod tání: 202…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006777 CAS číslo: 611-36-9 EINECS: 210-268-2 UN: 2811 ADR: 6.1,III |
Výsledky specifikačního testu
Appearance | Yellow to light beige Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 202 to 206 °C |
Titration with HClO4 | ≥97.5 % |
Specific optical rotation | +9.5° to +10.5° (20 °C, 589 nm) (c=4 in H2O) reading after >1 hour |
Iron (Fe) | ≤0.2 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...