8-Nitroquinoline ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
B23574.06
B23574.14
B23574.22
B23574.06EA
1190
CZK
B23574.06
B23574.14
B23574.22
8-Nitroquinoline ≥98%
8-Nitroquinoline
Purity: 98%
CAS No.: 607-35-2
Molecular Formula: C9H6N2O2
Formula Weight: 174.16
Melting Point: 86-90°
Beilstein Reference: 135178
Vzorec:
C₉H₆N₂O₂ MW: 174,16 g/mol Bod varu: 323 °C (1013 hPa) Bod tání: 86…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006806 CAS číslo: 607-35-2 EINECS: 210-135-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...