3-Quinolinecarbaldehyde ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Quinoline-3-carboxaldehyde, Quinoline-3-carbaldehyde, 3-Quinolinecarboxaldehyde
L13815.03
L13815.06
L13815.MD
L13815.03EA
3950
CZK
L13815.03
L13815.06
L13815.MD
3-Quinolinecarbaldehyde ≥98%
3-Quinolinecarbaldehyde
Purity: 98+%
CAS No.: 13669-42-6
Molecular Formula: C10H7NO
Formula Weight: 157.17
Melting Point: 68-71°
Beilstein Reference: 113269
Vzorec:
C₁₀H₇NO MW: 157,17 g/mol Bod tání: 68…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006768 CAS číslo: 13669-42-6 EINECS: 237-147-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...