(1S)-(-)-α-Pinene mixture of enantiomers ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
(-)-α-Pinene, (1S)-(-)-2-Pinene, (-)-Pin-2(3)-ene, L(-)-α-Pinene
L04759.18
L04759.30
L04759.18EA
790
CZK
L04759.18
L04759.30
(1S)-(-)-α-Pinene mixture of enantiomers ≥98%
(1S)-(-)-α-Pinene
Purity: 98%
CAS No.: 7785-26-4
Molecular Formula: C10H16
Formula Weight: 136.24
Flash Point: 32°(89°F)
Refractive Index: 1.4650
Melting Point: -64°
Boiling Point: 154-156°
Beilstein Reference: 1903790
Merck Reference: 14,7445
Optical Rotation: -42° (neat)
UN No.: 2368
MDL No.: MFCD00064145
Vzorec:
C₁₀H₁₆ MW: 136.24 Bod varu: 154…156 °C Bod tání: –64 °C Hustota: 0.8592 Bod vzplanutí: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064145 CAS číslo: 7785-26-4 EINECS: 232-077-3 UN: 2368 ADR: 3 III Merck Index: 14,7445 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...