1,3,5-Triphenylbenzene ≥99%
Dodavatel: Thermo Scientific
B21353.0B
B21353.18
B21353.30
B21353.0BEA
22620
CZK
B21353.0B
B21353.18
B21353.30
1,3,5-Triphenylbenzene ≥99%
1,3,5-Triphenylbenzene
Purity: 99+%
CAS No.: 612-71-5
Molecular Formula: C24H18
Formula Weight: 306.41
Melting Point: 172-174°
Boiling Point: 460°
MDL No.: MFCD00003060
Vzorec:
C₂₄H₁₈ MW: 306,41 g/mol Bod varu: 460 °C Bod tání: 172…174 °C Hustota: 1,22 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003060 CAS číslo: 612-71-5 EINECS: 210-318-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...