Tisk…

2-Phenylcyclohexanone 98

Dodavatel: Thermo Fisher Scientific

A17340.03 A17340.06 A17340.14
A17340.03EA 1330 CZK
A17340.03 A17340.06 A17340.14
2-Phenylcyclohexanone 98
2-Phenylcyclohexanone

Purity: 98% CAS No.: 1444-65-1 Molecular Formula: C12H14O Formula Weight: 174.24 Melting Point: 56-59° MDL No.: MFCD00001629

Vzorec: C₁₂H₁₄O
MW: 174.24
Bod varu: 90 °C (0.5 mmHg)
Bod tání: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00001629
CAS číslo: 1444-65-1
EINECS: 215-888-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR