Tisk…

3-Bromo-1,2,4,5-tetramethylbenzene ≥99%

Dodavatel: Thermo Scientific
Varování

L01465.06 L01465.14
L01465.06EA 2560 CZK
L01465.06 L01465.14
3-Bromo-1,2,4,5-tetramethylbenzene ≥99%
3-Bromo-1,2,4,5-tetramethylbenzene

Purity: 99% CAS No.: 1646-53-3 Molecular Formula: C10H13Br Formula Weight: 213.12 Melting Point: 58-62° Boiling Point: 115-120°/5mm MDL No.: MFCD00000070

Vzorec: C₁₀H₁₃Br
Bod varu: 115…120 °C (5 mmHg)
Bod tání: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00000070
CAS číslo: 1646-53-3
EINECS: 216-707-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR